accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNONCH3HNOHONO

CAS RN 1173900-33-8

IUPAC Name: 2-({1-[7-methyl-2-(morpholin-4-yl)-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl]ethyl}amino)benzoic acid
Synonyms: 2-({1-[7-methyl-2-(morpholin-4-yl)-4-oxopyrido[1,2-a]pyrimidin-9-yl]ethyl}amino)benzoic acid
Molecular Weight (g/mol): 408.46
Molecular Formula: C22H24N4O4
InChi Key: IRTDIKMSKMREGO-UHFFFAOYNA-N
SMILES: CC(NC1=CC=CC=C1C(O)=O)C1=CC(C)=CN2C(=O)C=C(N=C12)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results