accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO(R)NHNPOHOHO(E)

CAS RN 117414-74-1

IUPAC Name: (2R)-4-[(2E)-3-phosphonoprop-2-en-1-yl]piperazine-2-carboxylic acid
Synonyms: midafotel
Molecular Weight (g/mol): 250.19
Molecular Formula: C8H15N2O5P
InChi Key: VZXMZMJSGLFKQI-ABVWVHJUSA-N
SMILES: OC(=O)[C@H]1CN(C\C=C\P(O)(O)=O)CCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results