accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONOH

CAS RN 1177356-70-5

IUPAC Name: 3a-hydroxy-6-methyl-1-phenyl-1H,2H,3H,3aH,4H-pyrrolo[2,3-b]quinolin-4-one
Synonyms: 3a-hydroxy-6-methyl-1-phenyl-2H,3H-pyrrolo[2,3-b]quinolin-4-one
Molecular Weight (g/mol): 292.34
Molecular Formula: C18H16N2O2
InChi Key: LZAXPYOBKSJSEX-UHFFFAOYNA-N
SMILES: CC1=CC2=C(C=C1)N=C1N(CCC1(O)C2=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results