Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-Blebbistatin, Tocris Bioscience™

Catalog No. 18531
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-531 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-531 Supplier R&D Systems Supplier No. 1853/1
This item is not returnable. View return policy

Selective inhibitor of myosin II ATPase activity. Inactive enantiomer of (±)-blebbistatin (Cat. No. 1760)

Inactive enantiomer of the selective inhibitor of myosin II (±)-blebbistatin.

Active Enantiomer and Racemate also available.

Chemical Identifiers

CAS 1177356-70-5
Molecular Formula C18H16N2O2
Molecular Weight (g/mol) 292.338
InChI Key LZAXPYOBKSJSEX-SFHVURJKSA-N
Synonym +-blebbistatin, r-+-blebbistatin, blebbistatin, +, unii-xwg1958e8i, insolutionblebbistatin, racemic, r-blebbistatin, +-brebbistatin, tocris-1760, 4h-pyrrolo 2,3-b quinolin-4-one, 1,2,3,3a-tetrahydro-3a-hydroxy-6-methyl-1-phenyl-, 3ar, unii-20wc4j7cq6 component
PubChem CID 6604910
ChEBI CHEBI:75387
IUPAC Name (3aR)-3a-hydroxy-6-methyl-1-phenyl-2,3-dihydropyrrolo[2,3-b]quinolin-4-one
SMILES CC1=CC2=C(C=C1)N=C3C(C2=O)(CCN3C4=CC=CC=C4)O

Specifications

Quantity 1 mg
Formula Weight 292.34
Percent Purity >99%
Chemical Name or Material (R)-(+)-Blebbistatin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.