accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHSOOBrOCH3

CAS RN 1179183-69-7

IUPAC Name: 2-bromo-N-(3,4-dimethoxyphenyl)benzene-1-sulfonamide
Synonyms: 2-bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide
Molecular Weight (g/mol): 372.23
Molecular Formula: C14H14BrNO4S
InChi Key: WPZXLTFEOQPBPJ-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(NS(=O)(=O)C2=CC=CC=C2Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results