accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONSOOHOONSOOONa

CAS RN 117961-20-3

IUPAC Name: sodium 3-(morpholin-4-yl)propane-1-sulfonic acid 3-(morpholin-4-yl)propane-1-sulfonate
Synonyms: sodium morpholinopropanesulfonic acid 3-(morpholin-4-yl)propane-1-sulfonate
Molecular Weight (g/mol): 440.5
Molecular Formula: C14H29N2NaO8S2
InChi Key: OVCWZKQPQOVIDT-UHFFFAOYSA-M
SMILES: [Na+].OS(=O)(=O)CCCN1CCOCC1.[O-]S(=O)(=O)CCCN1CCOCC1
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)