accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOONOONOO

CAS RN 118-96-7

IUPAC Name: 2-methyl-1,3,5-trinitrobenzene
Synonyms: trinitrotoluene trotyl TNT trinitrotoluen trinitrotoluol sym-trinitrotoluol α-tnt α-tnt tritol s-trinitrotoluol
Molecular Weight (g/mol): 227.13
Molecular Formula: C7H5N3O6
InChi Key: SPSSULHKWOKEEL-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
Solvent or Matrix 
  • (1)
  • (1)
Element 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)