Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trinitrotoluene Solution, 1000μg/mL in acetonitrile, Ultra Scientific

Catalog No. USEPA1243
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1243 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1243 Supplier Agilent Technologies Supplier No. EPA12431
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 118-96-7 , 75-05-8
Molecular Formula C7H5N3O6
Molecular Weight (g/mol) 227.132
InChI Key SPSSULHKWOKEEL-UHFFFAOYSA-N
PubChem CID 8376
ChEBI CHEBI:46053
IUPAC Name 2-methyl-1,3,5-trinitrobenzene
SMILES CC1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-]

Specifications

CAS Max % 99.87, 0.12
Analyte or Component Names tert-butylmethyl ether (MTBE),trichloroacetic acid
Element Tert-butylmethyl ether, Trichloroacetic Acid
Solvent or Matrix MTBE
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material 2, 4, 6-Trinitrotoluene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.