CAS RN 118372-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ JAK2 Inhibitor III, SD-1029
The JAK2 Inhibitor III, SD-1029, also referenced under CAS 118372-34-2, controls the biological activity of JAK2.