accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1184-57-2

IUPAC Name: methylmercurylium hydroxide
Synonyms: methyl mercury ion hydroxide
Molecular Weight (g/mol): 232.63
Molecular Formula: CH4HgO
InChi Key: KRZWEBVPFGCYMY-UHFFFAOYSA-M
SMILES: [OH-].C[Hg+]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results