accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1184-85-6

IUPAC Name: N-methylmethanesulfonamide
Synonyms: N-methylmethanesulfonamide
Molecular Weight (g/mol): 109.14
Molecular Formula: C2H7NO2S
InChi Key: UHNHTTIUNATJKL-UHFFFAOYSA-N
SMILES: CNS(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results