Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylmethanesulfonamide 98.0+%, TCI America™
SDP

Catalog No. M217025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M217025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M217025G Supplier TCI America Supplier No. M217025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1184-85-6
Molecular Formula C2H7NO2S
Molecular Weight (g/mol) 109.143
MDL Number MFCD00776277
InChI Key UHNHTTIUNATJKL-UHFFFAOYSA-N
PubChem CID 97632
IUPAC Name N-methylmethanesulfonamide
SMILES CNS(=O)(=O)C

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 g
UN Number 1993
Formula Weight 109.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methylmethanesulfonamide
RTECSNumber PB0476750
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.