CAS RN 1185-57-5
Ammonium iron(III) citrate, 16.5 to 18.5% Fe
CAS: 1185-57-5 Molecular Formula: C6H11FeNO7 Molecular Weight (g/mol): 265 MDL Number: MFCD00013099 InChI Key: FRHBOQMZUOWXQL-UHFFFAOYSA-N Synonym: ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz PubChem CID: 118984355 IUPAC Name: azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
Ammonium iron(III) citrate, Thermo Scientific Chemicals
Chemical Name or Material: Ammonium iron(III) citrate Name Note: 16.5 to 18.5% Fe CAS: 1185-57-5 Molecular Formula: C6H11FeNO7+3 Formula Weight: 265 MDL Number: MFCD00013099 Synonym: ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz
Ferric ammonium citrate, MP Biomedicals™
CAS: 1185-57-5 Molecular Formula: C6H11FeNO7+3 Molecular Weight (g/mol): 264.999 InChI Key: FRHBOQMZUOWXQL-UHFFFAOYSA-N Synonym: ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz PubChem CID: 118984355 IUPAC Name: azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
Ammonium Ferric Citrate (Laboratory), Fisher Chemical
CAS: 1185-57-5 Molecular Formula: C6H11FeNO7+3 Molecular Weight (g/mol): 264.999 MDL Number: MFCD00013099 InChI Key: FRHBOQMZUOWXQL-UHFFFAOYSA-N Synonym: ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz PubChem CID: 118984355 IUPAC Name: azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
Ammonium iron(III) citrate, brown
CAS: 1185-57-5 Molecular Formula: C6H11FeNO7+3 Molecular Weight (g/mol): 264.999 MDL Number: MFCD00013099 InChI Key: FRHBOQMZUOWXQL-UHFFFAOYSA-N Synonym: ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz PubChem CID: 118984355 IUPAC Name: azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
Ammonium Iron(Iii) Citrate, Solstice
CAS: 1185-57-5 Molecular Formula: C6H11FeNO7+3 Molecular Weight (g/mol): 264.999 InChI Key: FRHBOQMZUOWXQL-UHFFFAOYSA-N Synonym: ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz PubChem CID: 118984355 IUPAC Name: azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
Fisher Science Education™ Ferric Ammonium Citrate
CAS: 1185-57-5 Molecular Formula: C6H11FeNO7+3 Molecular Weight (g/mol): 264.999 InChI Key: FRHBOQMZUOWXQL-UHFFFAOYSA-N Synonym: ammonium ferric citrate,ferri seltz,ammonii ferri citras,ferric ammonium citrate,iron ammonium citrate,ammonium iron 3+ citrate,ammonium iron iii citrate,iron ammonium citrate, green,ferri-ammoniumcitrat, braunes,ferriseltz PubChem CID: 118984355 IUPAC Name: azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
Ferric Ammonium Citrate, Green, Powder, FCC, 14.5-16%, Spectrum™ Chemical
CAS: 1185-57-5 Molecular Formula: C6H8FeNO7 Molecular Weight (g/mol): 261.98 InChI Key: FRHBOQMZUOWXQL-UHFFFAOYSA-K IUPAC Name: iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine SMILES: N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O