Learn More
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
| Color | Green |
| Physical Form | Powder |
| Quantity | 50 g |
| Packaging | Plastic Bottle |
| Linear Formula | C6H8O7 ·xFe3+ · yNH3 |
| Grade | purum p.a. |
| Concentration or Composition (by Analyte or Components) | 14.5 to 16% Fe basis (RT) |
| Chemical Name or Material | Ammonium Iron(Iii) Citrate |
Safety and Handling
| Hazard Statement | P261-P305 + P351 + P338 |
| Precautionary Statement | H315-H319-H335 |
| EINECSNumber | 214-686-6 |
| RTECSNumber | GE7540000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.