Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferric Ammonium Citrate
SCI_ED

Catalog No. S25315 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25315 500 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25315
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7+3
Molecular Weight (g/mol) 264.999
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Quantity 500 g
Assay Percent Range 100% w/v
Packaging Solid
Chemical Name or Material Ferric Ammonium Citrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.