CAS RN 1186195-59-4
IUPAC Name:
5-[4'-(3-{[3',6'-bis(dimethylamino)-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-5-yl]formamido}propyl)-[1,1'-biphenyl]-4-yl]-1-(2,4-dichlorophenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide
Synonyms:
5-(4'-{3-[3',6'-bis(dimethylamino)-3-oxospiro[2-benzofuran-1,9'-xanthen]-5-ylformamido]propyl}-[1,1'-biphenyl]-4-yl)-1-(2,4-dichlorophenyl)-4-methyl-N-(piperidin-1-yl)pyrazole-3-carboxamide
Molecular Weight (g/mol):
974.98
Molecular Formula:
C56H53Cl2N7O5
InChi Key:
RNAPRXIZPJDYPH-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC2=C(C=C1)C1(OC(=O)C3=C1C=CC(=C3)C(=O)NCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C1=C(C)C(=NN1C1=C(Cl)C=C(Cl)C=C1)C(=O)NN1CCCCC1)C1=C(O2)C=C(C=C1)N(C)C
1
–
1
of
1
results
1
R&D Systems™ Tocrifluor T1117
CAS: 1186195-59-4 Molecular Formula: C56H53Cl2N7O5