Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Tocrifluor T1117
SDP

Catalog No. 2540100U Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-401-00U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-401-00U Supplier R&D Systems™ Supplier No. 2540/100U

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Fluorescent cannabinoid ligand; fluorescent form of AM 251 (Cat. No. 1117)

Fluorescent form of AM 251 . AM 251 conjugated with 5-carboxytetramethylrhodamine (5-TAMRA) that fluoresces at 543 nm excitation (590 nm emission). Displays GPR55 binding activity.

View more information regarding Tocrifluor T1117 .

Chemical Identifiers

CAS 1186195-59-4
Molecular Formula C56H53Cl2N7O5

Specifications

Quantity 100 μg
Product Line Tocris
Formula Weight 974.97
Purity >95%
Molecular Weight (g/mol) 974.984
Chemical Name or Material Tocrifluor T1117
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.