accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOOSOOONa

CAS RN 1186195-63-0

IUPAC Name: sodium 1-(2-nitrophenyl)ethyl sulfate
Synonyms: sodium 1-(2-nitrophenyl)ethyl sulfate
Molecular Weight (g/mol): 269.2
Molecular Formula: C8H8NNaO6S
InChi Key: LCCXAXUWDTVOHF-UHFFFAOYNA-M
SMILES: [Na+].CC(OS([O-])(=O)=O)C1=CC=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results