accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHH3C

CAS RN 1187-58-2

IUPAC Name: N-methylpropanamide
Synonyms: N-methyl propionamide
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: QJQAMHYHNCADNR-UHFFFAOYSA-N
SMILES: CCC(=O)NC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results