accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1188-14-3

IUPAC Name: triethylgermane
Synonyms: triethylgermane
Molecular Weight (g/mol): 160.82
Molecular Formula: C6H16Ge
InChi Key: LTLGQZPDSQYFSB-UHFFFAOYSA-N
SMILES: CC[GeH](CC)CC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)