Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylgermanium hydride, 97%

Catalog No. AA4317606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4317606 5 g
AA4317603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4317606 Supplier Thermo Scientific Chemicals Supplier No. 04317606
Add to Cart

missing translation for 'validMainframeMsg'

Co-reactant for the preparation of germanium acrylates | CAS: 1188-14-3 | C6H16Ge | 160.82 g/mol

Co-reactant for the preparation of germanium acrylates. They are the Synthetic Reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188-14-3
Molecular Formula C6H16Ge
Molecular Weight (g/mol) 160.82
MDL Number MFCD00077989
InChI Key LTLGQZPDSQYFSB-UHFFFAOYSA-N
Synonym hydride, triethylgermyl, triethyl germanium hydride
PubChem CID 6327150
IUPAC Name triethylgermanium
SMILES CC[GeH](CC)CC

Specifications

Density 1.004 g/mL
Boiling Point 122°C
Flash Point 5°C (41°F)
Refractive Index 1.4382
Quantity 5 g
UN Number UN3399
Solubility Information Soluble in water( 46g/100 mL (26°C).
Formula Weight 160.78
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethylgermanium hydride
Show More Show Less
Hazard Category H225-H261-H315-H319-H335
Hazard Statement GHS H Statement
H225-H261-H315-H319-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.