accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNHOOHCl

CAS RN 1188890-36-9

IUPAC Name: N-[4-(morpholin-4-yl)butyl]-1-benzofuran-2-carboxamide hydrochloride
Synonyms: N-[4-(morpholin-4-yl)butyl]-1-benzofuran-2-carboxamide hydrochloride
Molecular Weight (g/mol): 338.83
Molecular Formula: C17H23ClN2O3
InChi Key: PIOACXKZWXHBRB-UHFFFAOYSA-N
SMILES: Cl.O=C(NCCCCN1CCOCC1)C1=CC2=CC=CC=C2O1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results