accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrNH2HCl

CAS RN 1189924-80-8

IUPAC Name: 1-(3-bromo-5-fluorophenyl)methanamine hydrochloride
Synonyms: 1-(3-bromo-5-fluorophenyl)methanamine hydrochloride
Molecular Weight (g/mol): 240.5
Molecular Formula: C7H8BrClFN
InChi Key: OFYIDRUSNSFKEG-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC(F)=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results