accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1190332-25-2

IUPAC Name: 2-{4-[5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolane-2-carbonyl]piperazin-1-yl}-N-(1-oxo-2,3-dihydro-1H-isoindol-4-yl)acetamide dihydrochloride
Synonyms: 2-{4-[5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolane-2-carbonyl]piperazin-1-yl}-N-(1-oxo-2,3-dihydroisoindol-4-yl)acetamide dihydrochloride
Molecular Weight (g/mol): 610.45
Molecular Formula: C24H29Cl2N9O6
InChi Key: VVMQSDIMNDTMII-UHFFFAOYNA-N
SMILES: Cl.Cl.NC1=C2N=CN(C3OC(C(O)C3O)C(=O)N3CCN(CC(=O)NC4=CC=CC5=C4CNC5=O)CC3)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results