CAS RN 1191237-69-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ GS 441524
Viral RNA-dependent RNA polymerase (RdRP) inhibitor; broad spectrum antiviral nucleotide