CAS RN 119139-23-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Bisindolylmaleimide IV
A potent, cell permeable, and selective inhibitor of protein kinase C (IC50 = 87 nM).