accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOS(E)

CAS RN 1195-52-4

IUPAC Name: (2E)-3-(thiophen-3-yl)prop-2-enoate
Synonyms: (2E)-3-(thiophen-3-yl)prop-2-enoate
Molecular Weight (g/mol): 153.18
Molecular Formula: C7H5O2S
InChi Key: VYRYYUKILKRGDN-OWOJBTEDSA-M
SMILES: [O-]C(=O)\C=C\C1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results