accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3OOHNHO(S)CH3(S)CH3(R)ClCl

CAS RN 119509-24-9

IUPAC Name: 3-[(2S,4S,5R)-5,6-dichloro-2,4-dimethylhexanoyl]-2-hydroxy-5,6-dimethoxy-1,4-dihydropyridin-4-one
Synonyms: atpenin A5
Molecular Weight (g/mol): 366.24
Molecular Formula: C15H21Cl2NO5
InChi Key: OVULNOOPECCZRG-CIUDSAMLSA-N
SMILES: COC1=C(OC)C(=O)C(C(=O)[C@@H](C)C[C@H](C)[C@@H](Cl)CCl)=C(O)N1
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results