Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atpenin A5, MP Biomedicals™

Catalog No. MP218335701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218335701 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
UN3462
Catalog No. MP218335701 Supplier MP Biomedicals, Inc Supplier No. 0218335701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UN3462

Mitochondrial Complex II (succinate dehydrogenase or succinate:ubiquinone oxidoreductase) couples the oxidation of succinate to fumarate with the reduction of quinone to quinol. It is a member of the Krebs cycle.

  • Highly selective compound, ubiquinone-binding site mitochondrial complex II inhibitor, stimulates mitochondrial channels
  • Shows anti-ischemic and cardioprotective effects in vivo
  • Used in the Krebs cycle to fumarate with the reduction of quinone to quinol
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119509-24-9
Molecular Formula C15H21Cl2NO5
Synonym FO 125A5, atpenin A5, Antibiotic FO 125A5, Atpenin A5 (Synthetic), (2 S,4 S,5 R)-5,6-Dichloro-1-(2,4-dihydroxy-5,6-dimethoxypyridin-3-yl)-2,4-dimethyl-1-hexanone, (2 S,4 S,5 R)-1-(2,4-Dihydroxy-5,6-dimethoxy-3-pyridinyl)-2,4-dimethyl-5,6-dichloro-1-hexanone, 3-[(2 S,4 S,5 R)-5,6-Dichloro-2,4-dimethyl-1-oxohexyl]-4-hydroxy-5,6-dimethoxy-2(1 H)-pyridinone

Specifications

Quantity 1 mg
Melting Point 83°C to 85°C
Shipping Condition -20°C
Color White
Purity 96.1
Molecular Weight (g/mol) 366.2
Chemical Name or Material Atpenin A5
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.