accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONFFFONHN

CAS RN 1196109-52-0

IUPAC Name: N-(pyridin-3-yl)-4-[(3-{[5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)methyl]piperidine-1-carboxamide
Synonyms: N-(pyridin-3-yl)-4-[(3-{[5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)methyl]piperidine-1-carboxamide
Molecular Weight (g/mol): 456.47
Molecular Formula: C24H23F3N4O2
InChi Key: NBOJHRYUGLRASX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(OC2=CC=CC(CC3CCN(CC3)C(=O)NC3=CN=CC=C3)=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results