CAS RN 1197334-04-5
IUPAC Name:
5-methyl-N-(pyridin-3-yl)-1H,2H,3H,5H-pyrrolo[2,3-f]indole-1-carboxamide hydrochloride
Synonyms:
5-methyl-N-(pyridin-3-yl)-2H,3H-pyrrolo[2,3-f]indole-1-carboxamide hydrochloride
Molecular Weight (g/mol):
328.8
Molecular Formula:
C17H17ClN4O
InChi Key:
VGEMBOFBPSNOIO-UHFFFAOYSA-N
SMILES:
Cl.CN1C=CC2=CC3=C(CCN3C(=O)NC3=CN=CC=C3)C=C12
1
–
1
of
1
results
1
R&D Systems™ SB 206553 hydrochloride
CAS: 1197334-04-5 Molecular Formula: C17H17ClN4O