accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CCH3NHPHOOOOH3COHOHOHOHNNHOHO

CAS RN 119942-99-3

IUPAC Name: 2-[2-({hydroxy[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]phosphoryl}amino)-4-methylpentanamido]-3-(1H-indol-3-yl)propanoic acid sodium
Synonyms: phosphoramidon sodium
Molecular Weight (g/mol): 566.5
Molecular Formula: C23H34N3NaO10P
InChi Key: KITONROGOABYFZ-UHFFFAOYNA-N
SMILES: [Na].CC(C)CC(NP(O)(=O)OC1OC(C)C(O)C(O)C1O)C(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results