accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3H3CCH3OOON

CAS RN 1199576-10-7

IUPAC Name: 6'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-3'-oxa-17'-azaspiro[2-benzofuran-1,10'-pentacyclo[11.7.1.0²,¹¹.0⁴,⁹.0¹⁷,²¹]henicosane]-1'(21'),2'(11'),4'(9'),5',7',12'-hexaen-3-one
Synonyms: 6'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3'-oxa-17'-azaspiro[2-benzofuran-1,10'-pentacyclo[11.7.1.0²,¹¹.0⁴,⁹.0¹⁷,²¹]henicosane]-1'(21'),2'(11'),4'(9'),5',7',12'-hexaen-3-one
Molecular Weight (g/mol): 521.42
Molecular Formula: C32H32BNO5
InChi Key: KNTNXZHHCPNNGB-UHFFFAOYNA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1(OC(=O)C3=CC=CC=C13)C1=C(O2)C2=C3N(CCCC3=C1)CCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)