accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 120-08-1

IUPAC Name: 6,7-dimethoxy-2H-chromen-2-one
Synonyms: scoparone aesculetin dimethyl ether 6,7-dimethylesculetin escoparone 6,7-dimethoxy-2-benzopyrone aesculetin dimethylether scoparon esculetin dimethyl ether
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: GUAFOGOEJLSQBT-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C=CC(=O)OC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results