accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3NO

CAS RN 120-22-9

IUPAC Name: N,N-diethyl-4-nitrosoaniline
Synonyms: N,N-diethyl-4-nitrosoaniline
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: OLNMJIHADFYHAK-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)N=O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)