Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitroso-N,N-Diethylaniline, MP Biomedicals™

Catalog No. ICN15592750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15592750 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15592750 Supplier MP Biomedicals, Inc Supplier No. 0215592750

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-22-9
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
InChI Key OLNMJIHADFYHAK-UHFFFAOYSA-N
Synonym 4-nitroso-n,n-diethylaniline, n,n-diethyl-p-nitrosoaniline, diethylnitrosoaniline, p-nitrosodiethylaniline, benzenamine, n,n-diethyl-4-nitroso, 4-diethylaminonitrosobenzene, n,n-diethyl-4-nitrosobenzenamine, ccris 3087, aniline, n,n-diethyl-p-nitroso, p-nitroso-n,n-diethylaniline
PubChem CID 67115
IUPAC Name N,N-diethyl-4-nitrosoaniline
SMILES CCN(CC)C1=CC=C(C=C1)N=O

Specifications

Color Green
Quantity 50 g
Formula Weight 178.2
Physical Form Free-flowing Powder
Chemical Name or Material 4-Nitroso-N, N-Diethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.