Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-4-nitrosoaniline 98.0+%, TCI America™
SDP

Catalog No. N026025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N026025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N026025G Supplier TCI America Supplier No. N026025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-22-9
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00002064
InChI Key OLNMJIHADFYHAK-UHFFFAOYSA-N
Synonym 4-nitroso-n,n-diethylaniline, n,n-diethyl-p-nitrosoaniline, diethylnitrosoaniline, p-nitrosodiethylaniline, benzenamine, n,n-diethyl-4-nitroso, 4-diethylaminonitrosobenzene, n,n-diethyl-4-nitrosobenzenamine, ccris 3087, aniline, n,n-diethyl-p-nitroso, p-nitroso-n,n-diethylaniline
PubChem CID 67115
IUPAC Name N,N-diethyl-4-nitrosoaniline
SMILES CCN(CC)C1=CC=C(C=C1)N=O

Specifications

Melting Point 84°C
Quantity 25 g
UN Number 2926
Formula Weight 178.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diethyl-4-nitrosoaniline
RTECSNumber BX3610000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.