accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120-54-7

IUPAC Name: (piperidine-1-carbothioylsulfanyl)disulfanyl piperidine-1-carbodithioate
Synonyms: thiuram MT
Molecular Weight (g/mol): 384.67
Molecular Formula: C12H20N2S6
InChi Key: VNDRMZTXEFFQDR-UHFFFAOYSA-N
SMILES: S=C(SSSSC(=S)N1CCCCC1)N1CCCCC1
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)