Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentamethylenethiuram Tetrasulfide, MP Biomedicals

Catalog No. ICN213244 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213244 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213244 Supplier MP Biomedicals, Inc Supplier No. 0521324480

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-54-7
Molecular Formula C12H20N2S6
Molecular Weight (g/mol) 384.67
MDL Number MFCD00047474
InChI Key VNDRMZTXEFFQDR-UHFFFAOYSA-N
Synonym dipentamethylenethiuram tetrasulfide, thiuram mt, bis pentamethylene thiuram tetrasulfide, tetrone a, nocceler tra, noksera tra, usaf b-31, sanceler tra, tetron a, soxinol tra
PubChem CID 61049
IUPAC Name (piperidine-1-carbothioylsulfanyl)disulfanyl piperidine-1-carbodithioate
SMILES S=C(SSSSC(=S)N1CCCCC1)N1CCCCC1

Specifications

Quantity 100 g
Formula Weight 384.7
Chemical Name or Material Dipentamethylenethiuram Tetrasulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.