Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentamethylenethiuram Tetrasulfide (so called) [Vulcanization Accelerator], TCI America™
SDP

Catalog No. D027925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D027925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D027925G Supplier TCI America Supplier No. D027925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-54-7
Molecular Formula C12H20N2S6
Molecular Weight (g/mol) 384.67
MDL Number MFCD00047474
InChI Key VNDRMZTXEFFQDR-UHFFFAOYSA-N
Synonym dipentamethylenethiuram tetrasulfide, thiuram mt, bis pentamethylene thiuram tetrasulfide, tetrone a, nocceler tra, noksera tra, usaf b-31, sanceler tra, tetron a, soxinol tra
PubChem CID 61049
IUPAC Name (piperidine-1-carbothioylsulfanyl)disulfanyl piperidine-1-carbodithioate
SMILES S=C(SSSSC(=S)N1CCCCC1)N1CCCCC1

Specifications

Melting Point 120°C
Color Yellow
Quantity 25 g
Formula Weight 384.67
Physical Form Crystalline Powder
Chemical Name or Material Dipentamethylenethiuram Tetrasulfide (so called) [Vulcanization Accelerator]
EINECSNumber (5)-0793
RTECSNumber TN4221000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.