accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12013-82-0

IUPAC Name: calcium bis(calcioazanide)
Synonyms: calcium nitride
Molecular Weight (g/mol): 148.25
Molecular Formula: Ca3N2
InChi Key: XCNGEWCFFFJZJT-UHFFFAOYSA-N
SMILES: [Ca++].[N-]=[Ca].[N-]=[Ca]
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)