accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12027-47-3

IUPAC Name: tetrasodium silicic acid tetrakis(tetraoxotritungstoxan-1-oylolate)
Synonyms: tetrasodium silicic acid tetrakis(tetraoxotritungstoxan-1-oylolate)
Molecular Weight (g/mol): 2970.12
Molecular Formula: H4Na4O40SiW12
InChi Key: RFKXKZJSLAFLCE-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Na+].[Na+].O[Si](O)(O)O.[O-][W](=O)(=O)O[W](=O)(=O)O[W](=O)=O.[O-][W](=O)(=O)O[W](=O)(=O)O[W](=O)=O.[O-][W](=O)(=O)O[W](=O)(=O)O[W](=O)=O.[O-][W](=O)(=O)O[W](=O)(=O)O[W](=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results