accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNH2CH3OOHOOH(E/Z)

CAS RN 1204480-26-1

IUPAC Name: 10-(3-aminopropyl)-3,4-dimethyl-9,10-dihydroacridin-9-one; but-2-enedioic acid
Synonyms: 10-(3-aminopropyl)-3,4-dimethylacridin-9-one; butenedioic acid
Molecular Weight (g/mol): 396.44
Molecular Formula: C22H24N2O5
InChi Key: WVUQPGFRFBVJKH-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(O)=O.CC1=CC=C2C(=O)C3=CC=CC=C3N(CCCN)C2=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)