accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4FFFFFFNb

CAS RN 12062-13-4

IUPAC Name: niobium(5+) ammonium hexafluoride
Synonyms: niobium(5+) ammonium hexafluoride
Molecular Weight (g/mol): 224.94
Molecular Formula: F6H4NNb
InChi Key: SKIHFCFFRXCIJA-UHFFFAOYSA-I
SMILES: [NH4+].[F-].[F-].[F-].[F-].[F-].[F-].[Nb+5]
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results