Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium AA Standard, 1000 ppm Nb in H2O/tr HF, Ricca Chemical
SDP

Catalog No. ANB1KW500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ANB1KW500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ANB1KW500 Supplier Ricca Chemical Company Supplier No. ANB1KW500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12062-13-4 , 7664-39-3 , 7732-18-5

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.25, 0.1
CAS Max % 100.0, 0.27, 0.1
Odor Odorless
Color Colorless
Boiling Point 100°C
Type Standard
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg Nb (1, 000ppm Nb)NH4NbF6 in Water / tr HF
Chemical Name or Material Niobium AA Standard
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.