CAS RN 1206711-16-1
IUPAC Name:
4-{6-[4-(propan-2-yloxy)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl}quinoline
Synonyms:
4-[6-(4-isopropoxyphenyl)pyrazolo[1,5-a]pyrimidin-3-yl]quinoline
Molecular Weight (g/mol):
380.45
Molecular Formula:
C24H20N4O
InChi Key:
JMIFGARJSWXZSH-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=C2C=CC=CC2=NC=C1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ BMP Inhibitor II, DMH1
The BMP Inhibitor II, DMH1, also referenced under CAS 1206711-16-1, controls the biological activity of BMP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
2
R&D Systems™ DMH-1
CAS: 1206711-16-1 Molecular Formula: C24H20N4O