accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12070-14-3

IUPAC Name: methanidylidynezirconiumylium
Synonyms: methanidylidynezirconiumylium
Molecular Weight (g/mol): 103.24
Molecular Formula: CZr
InChi Key: MHMXYYDKIQTOHA-UHFFFAOYSA-N
SMILES: [C-]#[Zr+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results