accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12079-65-1

MnCOCOCOHC

CAS RN 12079-65-1

IUPAC Name: cyclopenta-2,4-dien-1-ide tris(methanidylidyneoxidanium) manganese
Synonyms: tris(carbon monoxide) cyclopentadienide manganese
Molecular Weight (g/mol): 204.06
Molecular Formula: C8H5MnO3
InChi Key: CENDTHIEZAWVHS-UHFFFAOYSA-N
SMILES: [Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH-]1C=CC=C1
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results