accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121-34-6

IUPAC Name: 4-hydroxy-3-methoxybenzoic acid
Synonyms: vanillic acid acide vanillique p hydroxy m methoxy benzoic acid p-vanillic acid 4 hydroxy 3 methoxybenzoic acid acid, vanillic p-vanillate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: WKOLLVMJNQIZCI-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1O)C(O)=O
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)