Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methoxybenzoic acid, 97+%

Catalog No. AC121351000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121351000 100 g Glass bottle
AC121350250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC121351000 Supplier Thermo Scientific Chemicals Supplier No. 121351000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 121-34-6 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-34-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002551
InChI Key WKOLLVMJNQIZCI-UHFFFAOYSA-N
Synonym vanillic acid, acide vanillique, p-vanillic acid, benzoic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxybenzoic acid, vanillate, m-anisic acid, 4-hydroxy, protocatechuic acid, 3-methyl ester, 4-hydroxy-3-methoxy-benzoic acid, unii-gm8q3jm2y8
PubChem CID 8468
ChEBI CHEBI:30816
IUPAC Name 4-hydroxy-3-methoxybenzoic acid
SMILES COC1=CC(=CC=C1O)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 210°C to 214°C
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CO2H
Quantity 100 g
Beilstein 10, 392
Merck Index 15, 10119
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol and ether
Formula Weight 168.15
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-3-methoxybenzoic acid, 98.5+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-466-8
RTECSNumber YW5300000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.